missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Pyridinedicarboxylic Acid 98%, ACROS Organics™

$39.40 - $39.40

Chemical Identifiers

CAS 100-26-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H5NO4
Linear Formula (C5H3N)(COOH)2
Formula Weight 167.12
MDL Number MFCD00006297
Synonym Isocinchomeronic acid
Chemical Name or Material 2, 5-Pyridinedicarboxylic acid, 98%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC131870050 SDS Thermo Fisher Scientific
131870050
Glass bottle 5g Each for $39.40
Specifications Description & Specifications

Chemical Identifiers

100-26-5
100.0
C7H5NO4
167.12
Isocinchomeronic acid
≥97.5%
97.5
98%
(C5H3N)(COOH)2
MFCD00006297
2, 5-Pyridinedicarboxylic acid, 98%

Specifications

167.12
5g
22, 153
100.0
Glass bottle
Authentic
98%
C7H5NO4
2, 5-Pyridinedicarboxylic acid, 98%
Fine Crystalline Powder
Yellow to green
15, 5204
Isocinchomeronic acid
97.5
100-26-5
(C5H3N)(COOH)2
MFCD00006297
≥97.5%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-834-2

TSCA : TSCA