missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Norbornadiene, 97%, stab with 250 ppm BHT

Manufacturer:  Alfa Aesar™ L13935AE

 View more versions of this product

Catalog No. AAL13935AE



Description

Description

Applications
2,5-Norbornadiene is used as an intermediate in prostaglandin synthesis and shows anti-ethylene effect on plants. It acts as a starting material for the synthesis of diamantane and as an acetylene transfer agent for instance, in reaction with 3,6-di-2-pyridyl-1,2,4,5-tetrazine. It is also a useful dienophile in Diels-Alder reactions.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications

Specifications

2,5-Norbornadiene
88°C to 90°C
100mL
121-46-0
MFCD00082301
506224
SJYNFBVQFBRSIB-UHFFFAOYSA-N
bicyclo[2.2.1]hepta-2,5-diene
8473
97%
−11°C (12°F)
-20°C
stabilized with 250ppm BHT
C7H8
UN2251
2,5-norbornadiene, bicyclo 2.2.1 hepta-2,5-diene, norbornadiene, unii-w9ztq75zus, bicyclo 2.2.1 hepta-2,5-diene, bicyclo 2.2.1 heptadiene, 8,9,10-trinorborna-2,5-diene, w9ztq75zus, bicyclo 2.2.1 heptadiene, dicycloheptadiene
C1C2C=CC1C=C2
92.141
92.14

Chemical Identifiers

121-46-0
92.141
SJYNFBVQFBRSIB-UHFFFAOYSA-N
8473
C1C2C=CC1C=C2
C7H8
MFCD00082301
2,5-norbornadiene, bicyclo 2.2.1 hepta-2,5-diene, norbornadiene, unii-w9ztq75zus, bicyclo 2.2.1 hepta-2,5-diene, bicyclo 2.2.1 heptadiene, 8,9,10-trinorborna-2,5-diene, w9ztq75zus, bicyclo 2.2.1 heptadiene, dicycloheptadiene
bicyclo[2.2.1]hepta-2,5-diene
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BICYCLO 2,2,1 HEPTA-2-5-DIENE, STABILIZED

RTECSNumber : RB6535000

TSCA : TSCA

Documents