Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(5-Nitro-2-pyridyloxy)ethanol, 98%

$117.84 - $117.84

Chemical Identifiers

Chemical Name or Material 2-(5-Nitro-2-pyridyloxy)ethanol
CAS 143071-39-0
Molecular Formula C7H8N2O4
Formula Weight 184.15
MDL Number MFCD00052642
Beilstein 150856
Synonym 2-(2-Hydroxyethoxy)-5-nitropyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0971303

  SDS
alfa aesar™
L0971303
1g Each for $117.84
Description & Specifications

Specifications

Formula Weight 184.15
Chemical Name or Material 2-(5-Nitro-2-pyridyloxy)ethanol
Assay 98%
Molecular Formula C7H8N2O4
MDL Number MFCD00052642
CAS 143071-39-0
Melting Point 112° to 114°C
Beilstein 150856
Synonym 2-(2-Hydroxyethoxy)-5-nitropyridine
Quantity 1g