missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-[(5-Nitro-2-pyridyl)oxy]ethan-1-ol, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CD08075EA
Specifications
2-[(5-Nitro-2-pyridyl)oxy]ethan-1-ol | |
96.5 | |
97% | |
C7H8N2O4 | |
10g | |
KESQFSZFUCZCEI-UHFFFAOYSA-N | |
2-(5-nitropyridin-2-yl)oxyethanol | |
587937 | |
97% |
143071-39-0 | |
100.0 | |
Amber Glass Bottle | |
MFCD00052642 | |
2-5-nitropyridin-2-yloxy ethanol, 2-5-nitro-2-pyridyloxy ethanol, 2-5-nitropyridin-2-yl oxy ethan-1-ol, 2-5-nitro-2-pyridyl oxy ethan-1-ol, 2-2-hydroxyethoxy-5-nitropyridine, 2-5-nitropyridin-2-yl oxy ethanol, 2-5-nitro-2-pyridyloxy ethan-1-ol, acmc-209xhf, maybridge1_002682, 2-5-nitropyridin-2-yl oxyethanol | |
C1=CC(=NC=C1[N+](=O)[O-])OCCO | |
184.151 | |
184.15 |
Chemical Identifiers
143071-39-0 | |
184.151 | |
KESQFSZFUCZCEI-UHFFFAOYSA-N | |
587937 | |
C1=CC(=NC=C1[N+](=O)[O-])OCCO |
C7H8N2O4 | |
MFCD00052642 | |
2-5-nitropyridin-2-yloxy ethanol, 2-5-nitro-2-pyridyloxy ethanol, 2-5-nitropyridin-2-yl oxy ethan-1-ol, 2-5-nitro-2-pyridyl oxy ethan-1-ol, 2-2-hydroxyethoxy-5-nitropyridine, 2-5-nitropyridin-2-yl oxy ethanol, 2-5-nitro-2-pyridyloxy ethan-1-ol, acmc-209xhf, maybridge1_002682, 2-5-nitropyridin-2-yl oxyethanol | |
2-(5-nitropyridin-2-yl)oxyethanol |