Please login to your online account to display your discounted pricing

2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)ethan-1-ol, 97%, Maybridge


Quantity 25g

Manufacturer: fisher scientific company  BTB09930EB

 View more versions of this product

Catalog No. BTB09930EB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 203.24
Quantity 25g
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C12H13NO2
Chemical Name or Material 2-(5-Methyl-2-phenyl-1, 3-oxazol-4-yl)ethan-1-ol
MDL Number MFCD00100006
CAS 103788-65-4
CAS Min % 96.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-(5-Methyl-2-phenyl-1, 3-oxazol-4-yl)ethan-1-ol
CAS 103788-65-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H13NO2
Formula Weight 203.24
MDL Number MFCD00100006
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.