missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Lutidine, TCI America™

Manufacturer:  TCI America L006525ML

 View more versions of this product

Catalog No. L006525ML



Specifications

Specifications

2,5-Lutidine
Yellow
25mL
C7H9N
1993
XWKFPIODWVPXLX-UHFFFAOYSA-N
2,5-dimethylpyridine
11526
Clear Liquid at 20°C
157°C
-6°C
589-93-5
MFCD00006343
2,5-Dimethylpyridine
CC1=CN=C(C=C1)C
107.156
107.16
≥98.0% (GC)

Chemical Identifiers

589-93-5
107.156
XWKFPIODWVPXLX-UHFFFAOYSA-N
11526
CC1=CN=C(C=C1)C
C7H9N
MFCD00006343
2,5-Dimethylpyridine
2,5-dimethylpyridine
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0712

RTECSNumber : OK9625000

TSCA : Yes

Documents