Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(5-Isoxazolyl)phenol, 98%

$31.70 - $91.73

Chemical Identifiers

Chemical Name or Material 2-(5-Isoxazolyl)phenol
CAS 61348-47-8
Molecular Formula C9H7NO2
Formula Weight 161.16
MDL Number MFCD00218653
Synonym 5-(2-Hydroxyphenyl)isoxazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2749003

  SDS
alfa aesar™
H2749003
1g Each for $31.70

AAH2749006

  SDS
alfa aesar™
H2749006
5g Each for $91.73
Description & Specifications

Specifications

Formula Weight 161.16
Molecular Formula C9H7NO2
Chemical Name or Material 2-(5-Isoxazolyl)phenol
Assay 98%
MDL Number MFCD00218653
CAS 61348-47-8
Synonym 5-(2-Hydroxyphenyl)isoxazole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 183° to 184°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.