missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Hexanediol 98.0+%, TCI America™
Supplier: TCI America H010025ML
Specifications
| 2,5-Hexanediol | |
| Yellow | |
| C6H14O2 | |
| 25 mL | |
| OHMBHFSEKCCCBW-UHFFFAOYSA-N | |
| hexane-2,5-diol | |
| 18049 | |
| 118.18 | |
| Liquid |
| 2935-44-6 | |
| 221°C | |
| MFCD00004557 | |
| 2,5-hexanediol, diisopropanol, 2,5-dihydroxyhexane, 2,5-hexyleneglycol, 2,5-hexanediol, mixture of isomers, 2,5-hexandiol, hexan-2,5-diol, 2,5-hexylene glycol, acmc-1acz0, acmc-209e5m | |
| CC(CCC(C)O)O | |
| 118.176 | |
| CHEBI:84894 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2935-44-6 | |
| 118.176 | |
| OHMBHFSEKCCCBW-UHFFFAOYSA-N | |
| 18049 | |
| hexane-2,5-diol |
| C6H14O2 | |
| MFCD00004557 | |
| 2,5-hexanediol, diisopropanol, 2,5-dihydroxyhexane, 2,5-hexyleneglycol, 2,5-hexanediol, mixture of isomers, 2,5-hexandiol, hexan-2,5-diol, 2,5-hexylene glycol, acmc-1acz0, acmc-209e5m | |
| CHEBI:84894 | |
| CC(CCC(C)O)O |
Safety and Handling
EINECSNumber : (2)-0240
RTECSNumber : MO2275000
TSCA : Yes