Learn More
2,5-Dithiobiurea, 97%, Thermo Scientific Chemicals
$71.63 - $194.40
Chemical Identifiers
CAS | 142-46-1 |
---|---|
Molecular Formula | C2H6N4S2 |
Molecular Weight (g/mol) | 150.22 |
MDL Number | MFCD00014471 |
InChI Key | KCOYHFNCTWXETP-UHFFFAOYSA-N |
Synonym | 2,5-dithiobiurea, dithiobiurea, bithiourea, dithiourea, 1,2-hydrazinedicarbothioamide, bis thiourea, hydrazodicarbothioamide, 2,5-dithiodiurea, hydrazine-1,2-bis carbothioamide, dithiocarbamoylhydrazine |
PubChem CID | 2724564 |
IUPAC Name | (carbamothioylamino)thiourea |
SMILES | NC(=S)NNC(N)=S |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1666714
|
Thermo Scientific Chemicals
A1666714 |
25 g |
Each for $71.63
|
|
|||||
AAA1666722
|
Thermo Scientific Chemicals
A1666722 |
100 g |
Each for $194.40
|
|
|||||
Description
2,5-Dithiobiurea is used in radiopharmaceutical imaging, inhibiting enzyme function, kidney function study and to treat toxic metal poisoning. It is also used in co-crystals development and in the field of nonlinear optics to generate new coherent wavelengths
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,5-Dithiobiurea is used in radiopharmaceutical imaging, inhibiting enzyme function, kidney function study and to treat toxic metal poisoning. It is also used in co-crystals development and in the field of nonlinear optics to generate new coherent wavelengths
Solubility
Soluble in hot water, pyridine, triethylamine and dimethylformamide. Insoluble in cold water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
142-46-1 | |
150.22 | |
KCOYHFNCTWXETP-UHFFFAOYSA-N | |
2724564 | |
NC(=S)NNC(N)=S |
C2H6N4S2 | |
MFCD00014471 | |
2,5-dithiobiurea, dithiobiurea, bithiourea, dithiourea, 1,2-hydrazinedicarbothioamide, bis thiourea, hydrazodicarbothioamide, 2,5-dithiodiurea, hydrazine-1,2-bis carbothioamide, dithiocarbamoylhydrazine | |
(carbamothioylamino)thiourea |
Specifications
142-46-1 | |
C2H6N4S2 | |
25 g | |
2,5-dithiobiurea, dithiobiurea, bithiourea, dithiourea, 1,2-hydrazinedicarbothioamide, bis thiourea, hydrazodicarbothioamide, 2,5-dithiodiurea, hydrazine-1,2-bis carbothioamide, dithiocarbamoylhydrazine | |
KCOYHFNCTWXETP-UHFFFAOYSA-N | |
(carbamothioylamino)thiourea | |
2724564 | |
97% |
∼212°C (decomposition) | |
MFCD00014471 | |
1723178 | |
Soluble in hot water,pyridine,triethylamine and dimethylformamide. Insoluble in cold water. | |
NC(=S)NNC(N)=S | |
150.22 | |
150.22 | |
2,5-Dithiobiurea |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-537-6
RTECSNumber : EC1460000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only