missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,5-Diphenyloxazole, Scintillation Grade 99%, Thermo Scientific™
Chemical Identifiers
CAS | 92-71-7 |
---|---|
Molecular Formula | C15H11NO |
Molecular Weight (g/mol) | 221.26 |
MDL Number | MFCD00005306 |
InChI Key | CNRNYORZJGVOSY-UHFFFAOYSA-N |
Synonym | PPO |
PubChem CID | 7105 |
IUPAC Name | 2,5-diphenyl-1,3-oxazole |
SMILES | O1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC117380250
|
Thermo Scientific Chemicals
117380250 |
25g | Plastic bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC117381000
|
Thermo Scientific Chemicals
117381000 |
100g | Plastic bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC117385000
|
Thermo Scientific Chemicals
117385000 |
500g | Plastic bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC117380025
|
Thermo Scientific Chemicals
117380025 |
2.5kg | Plastic Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for fluorescence/luminescence spectroscopyChemical Identifiers
92-71-7 | |
221.26 | |
CNRNYORZJGVOSY-UHFFFAOYSA-N | |
7105 | |
O1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1 |
C15H11NO | |
MFCD00005306 | |
PPO | |
2,5-diphenyl-1,3-oxazole |
Specifications
92-71-7 | |
100.0 | |
1.06g/mL | |
25g | |
Authentic | |
MFCD00005306 | |
Plastic bottle | |
CNRNYORZJGVOSY-UHFFFAOYSA-N | |
2,5-diphenyl-1,3-oxazole | |
7105 | |
≥98.5% |
98.5 | |
< 25 cpm | |
White | |
99% | |
C15H11NO | |
27, 78 | |
PPO | |
O1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1 | |
221.26 | |
221.26 | |
2, 5-Diphenyloxazole, 99% |
Safety and Handling
EINECSNumber : 202-181-3
RTECSNumber : RP6825000
TSCA : TSCA
RUO â Research Use Only