Please login to your online account to display your discounted pricing

2,5-Diphenyl-1,3,4-oxadiazole 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2,5-Diphenyl-1,3,4-oxadiazole
Name Note 97%
CAS 725-12-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C14H10N2O
Formula Weight 222.25
MDL Number MFCD00003102
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC117370050

  SDS
thermo fisher scientific
117370050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥96.0%
Formula Weight 222.25
Quantity 5g
Color White
EINECS Number 211-968-0
RTECS Number RO0802500
Melting Point 136° to 141°C
TSCA TSCA
Beilstein 27, 591
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
Molecular Formula C14H10N2O
CAS 725-12-2
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2,5-Diphenyl-1,3,4-oxadiazole
Assay Percent Range 97%
MDL Number MFCD00003102
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.