Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Diphenyl-1,3,4-oxadiazole, 97%

$34.49 - $401.76

Chemical Identifiers

Chemical Name or Material 2,5-Diphenyl-1,3,4-oxadiazole
CAS 725-12-2
Molecular Formula C14H10N2O
Formula Weight 222.25
MDL Number MFCD00003102
Beilstein 170385
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0276303

 
alfa aesar™
L0276303
1g Each for $34.49

AAL0276306

 
alfa aesar™
L0276306
5g Each for $103.76

AAL0276314

 
alfa aesar™
L0276314
25g Each for $401.76
Description & Specifications

Specifications

Formula Weight 222.25
Chemical Name or Material 2,5-Diphenyl-1,3,4-oxadiazole
Molecular Formula C14H10N2O
CAS 725-12-2
Beilstein 170385
Health Hazard 1 Danger
Quantity 1g
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Assay 97%
MDL Number MFCD00003102
Melting Point 138° to 141°C
Boiling Point 231°C (13mmHg)
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number RO0802500
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.