Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethylthiophenol, 98%

Chemical Identifiers

Chemical Name or Material 2,5-Dimethylthiophenol
CAS 4001-61-0
Molecular Formula C8H10S
Formula Weight 138.23
MDL Number MFCD00010020
UN Number UN2810
Beilstein 2040348
Synonym 2,5-Dimethylbenzenethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2416406

  SDS
alfa aesar™
B2416406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2416414

  SDS
alfa aesar™
B2416414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2416422

  SDS
alfa aesar™
B2416422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 138.23
Assay 98%
Flash Point 83°C (181°F)
CAS 4001-61-0
Quantity 5g
Beilstein 2040348
Chemical Name or Material 2,5-Dimethylthiophenol
MDL Number MFCD00010020
Odor Unpleasant
Boiling Point 126° to 127°C (50mmHg)
Molecular Formula C8H10S
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
UN Number UN2810
Synonym 2,5-Dimethylbenzenethiol
TSCA TSCA
Health Hazard 2 GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.