missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethylpyrrolidine, 97%, Maybridge™

Manufacturer:  Fisher Scientific Company AC11676CB

 View more versions of this product

Catalog No. AC11676CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 5-Dimethylpyrrolidine
Authentic
1.4290 to 1.4320 (20°C, 589nm)
250mg
96.5
97%
MFCD00005254
16,147
ZEBFPAXSQXIPNF-UHFFFAOYSA-N
2,5-dimethylpyrrolidine
96034
Liquid
0.81g/mL
Amber Glass Bottle
Yellow
3378-71-0
100.0
C6H13N
20, 102
2,5-dimethyl-pyrrolidine, 2,5-dimethylpyrrolidine, mixture of cis and trans, technical grade, 2rs,5rs-2,5-dimethylpyrrolidine, pyrrolidine, 2,5-dimethyl
CC1CCC(N1)C
99.177
99.18
≥97.5%

Chemical Identifiers

3378-71-0
99.177
ZEBFPAXSQXIPNF-UHFFFAOYSA-N
96034
CC1CCC(N1)C
C6H13N
MFCD00005254
2,5-dimethyl-pyrrolidine, 2,5-dimethylpyrrolidine, mixture of cis and trans, technical grade, 2rs,5rs-2,5-dimethylpyrrolidine, pyrrolidine, 2,5-dimethyl
2,5-dimethylpyrrolidine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents
Product Certifications