missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethylpyrrolidine, 97%, Maybridge

Chemical Identifiers

CAS 3378-71-0
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.177
MDL Number MFCD00005254
InChI Key ZEBFPAXSQXIPNF-UHFFFAOYSA-N
Synonym 2,5-dimethyl-pyrrolidine, pyrrolidine, 2,5-dimethyl, 2rs,5rs-2,5-dimethylpyrrolidine, 2,5-dimethylpyrrolidine, mixture of cis and trans, technical grade
PubChem CID 96034
IUPAC Name 2,5-dimethylpyrrolidine
SMILES CC1CCC(N1)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC11676CB SDS View Documents Fisher Scientific Company
AC11676CB
250mg Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC11676DA SDS View Documents Fisher Scientific Company
AC11676DA
1g Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

3378-71-0
99.177
ZEBFPAXSQXIPNF-UHFFFAOYSA-N
96034
CC1CCC(N1)C
C6H13N
MFCD00005254
2,5-dimethyl-pyrrolidine, pyrrolidine, 2,5-dimethyl, 2rs,5rs-2,5-dimethylpyrrolidine, 2,5-dimethylpyrrolidine, mixture of cis and trans, technical grade
2,5-dimethylpyrrolidine

Specifications

0.81g/mL
Amber Glass Bottle
Yellow
3378-71-0
100.0
C6H13N
20, 102
2,5-dimethyl-pyrrolidine, pyrrolidine, 2,5-dimethyl, 2rs,5rs-2,5-dimethylpyrrolidine, 2,5-dimethylpyrrolidine, mixture of cis and trans, technical grade
CC1CCC(N1)C
99.177
99.18
≥97.5%
Authentic
1.4290 to 1.4320 (20°C, 589nm)
250mg
96.5
97%
MFCD00005254
16,147
ZEBFPAXSQXIPNF-UHFFFAOYSA-N
2,5-dimethylpyrrolidine
96034
Liquid
2, 5-Dimethylpyrrolidine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger