missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethylphenylhydrazine hydrochloride, 98%

$39.88 - $335.57

Chemical Identifiers

CAS 56737-78-1
Molecular Formula C8H12N2·HCl
Formula Weight 172.66
MDL Number MFCD00013382
Beilstein 6118180
Chemical Name or Material 2,5-Dimethylphenylhydrazine hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1249606 SDS Alfa Aesar™
L1249606
5g Each for $39.88
AAL1249622 SDS Alfa Aesar™
L1249622
100g Each for $335.57
AAL1249614 SDS Alfa Aesar™
L1249614
25g Each for $109.59
Specifications Description & Specifications

Chemical Identifiers

56737-78-1
172.66
6118180
C8H12N2·HCl
MFCD00013382
2,5-Dimethylphenylhydrazine hydrochloride

Specifications

56737-78-1
≈205°C
MFCD00013382
5g
98%
C8H12N2·HCl
172.66
6118180
2,5-Dimethylphenylhydrazine hydrochloride

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger