missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethylphenylhydrazine hydrochloride, 98%

Manufacturer:  Alfa Aesar™ L1249622

 View more versions of this product

Catalog No. AAL1249622



Specifications

Specifications

2,5-Dimethylphenylhydrazine hydrochloride
100g
C8H13ClN2
6118180
OMOFMSZLYCEIAF-UHFFFAOYSA-N
(2,5-dimethylphenyl)hydrazine;hydrochloride
2734058
98%
~205°C
56737-78-1
MFCD00013382
2,5-dimethylphenylhydrazine hydrochloride, 2,5-dimethylphenyl hydrazine hydrochloride, 2,5-dimethylphenylhydrazinehydrochloride, 2,3-dimethylphenylhydrazinehydrochloride, pubchem6400, acmc-1bf0t, ksc491e8b, 2-hydrazino-m-xylene hydrochloride, 2,5-dimethyl-phenylhydrazine hydrochloride, 1-2,5-dimethylphenyl hydrazine hydrochloride
CC1=CC(=C(C=C1)C)NN.Cl
172.656
172.66

Chemical Identifiers

56737-78-1
172.656
OMOFMSZLYCEIAF-UHFFFAOYSA-N
2734058
CC1=CC(=C(C=C1)C)NN.Cl
C8H13ClN2
MFCD00013382
2,5-dimethylphenylhydrazine hydrochloride, 2,5-dimethylphenyl hydrazine hydrochloride, 2,5-dimethylphenylhydrazinehydrochloride, 2,3-dimethylphenylhydrazinehydrochloride, pubchem6400, acmc-1bf0t, ksc491e8b, 2-hydrazino-m-xylene hydrochloride, 2,5-dimethyl-phenylhydrazine hydrochloride, 1-2,5-dimethylphenyl hydrazine hydrochloride
(2,5-dimethylphenyl)hydrazine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents