Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethylphenylacetonitrile, 98+%

Chemical Identifiers

Chemical Name or Material 2,5-Dimethylphenylacetonitrile
CAS 16213-85-7
Molecular Formula C10H11N
Formula Weight 145.2
MDL Number MFCD00013816
UN Number UN3439
Beilstein 2084136
Synonym 2,5-Dimethylbenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0194206

 
alfa aesar™
L0194206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0194214

 
alfa aesar™
L0194214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 145.2
Chemical Name or Material 2,5-Dimethylphenylacetonitrile
Molecular Formula C10H11N
Assay ≥98%
Boiling Point 73°C (1mmHg)
MDL Number MFCD00013816
CAS 16213-85-7
Beilstein 2084136
Quantity 5g
Synonym 2,5-Dimethylbenzyl cyanide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
RTECS Number AM2675000
Melting Point 27° to 29°C
UN Number UN3439
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.