Learn More
2,5-Dimethylphenylacetonitrile, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L0194214
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2,5-Dimethylphenylacetonitrile | |
27°C to 29°C | |
C10H11N | |
25 g | |
2084136 | |
HCELORQTHSKCQL-UHFFFAOYSA-N | |
2-(2,5-dimethylphenyl)acetonitrile | |
27753 | |
≥98% |
16213-85-7 | |
73°C (1 mmHg) | |
MFCD00013816 | |
UN3439 | |
2,5-dimethylphenylacetonitrile, 2-2,5-dimethylphenyl acetonitrile, 2,5-dimethylbenzyl cyanide, acetonitrile, 2,5-xylyl, 2,5-xylyl acetonitrile, benzeneacetonitrile, 2,5-dimethyl, 2,5-xylylacetonitrile, 2,5-dimethylphenyl acetonitrile, 2,5-dimethylbenzeneacetonitrile, unii-ult00p22mn | |
CC1=CC(=C(C=C1)C)CC#N | |
145.205 | |
145.2 |
Chemical Identifiers
16213-85-7 | |
145.205 | |
HCELORQTHSKCQL-UHFFFAOYSA-N | |
27753 | |
CC1=CC(=C(C=C1)C)CC#N |
C10H11N | |
MFCD00013816 | |
2,5-dimethylphenylacetonitrile, 2-2,5-dimethylphenyl acetonitrile, 2,5-dimethylbenzyl cyanide, acetonitrile, 2,5-xylyl, 2,5-xylyl acetonitrile, benzeneacetonitrile, 2,5-dimethyl, 2,5-xylylacetonitrile, 2,5-dimethylphenyl acetonitrile, 2,5-dimethylbenzeneacetonitrile, unii-ult00p22mn | |
2-(2,5-dimethylphenyl)acetonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 240-339-3
RTECSNumber : AM2675000
TSCA : No
Recommended Storage : Ambient temperatures