missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethylphenylacetone, 99+%, ACROS Organics™

Chemical Identifiers

CAS 53291-89-7
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.232
MDL Number MFCD00210423
InChI Key XHEUPKZIPXSSLU-UHFFFAOYSA-N
Synonym 1-2,5-dimethylphenyl acetone
PubChem CID 2734104
IUPAC Name 1-(2,5-dimethylphenyl)propan-2-one
SMILES CC1=CC(=C(C=C1)C)CC(=O)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC350890050 SDS View Documents Thermo Fisher Scientific
AC350890050
5g Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

53291-89-7
162.232
XHEUPKZIPXSSLU-UHFFFAOYSA-N
2734104
CC1=CC(=C(C=C1)C)CC(=O)C
C11H14O
MFCD00210423
1-2,5-dimethylphenyl acetone
1-(2,5-dimethylphenyl)propan-2-one

Specifications

Authentic
1.5170 to 1.5190 (20°C, 589nm)
5g
99.0
99+%
MFCD00210423
XHEUPKZIPXSSLU-UHFFFAOYSA-N
1-(2,5-dimethylphenyl)propan-2-one
2734104
Liquid
2, 5-Dimethylphenylacetone
Glass Bottle
Yellow
53291-89-7
100.0
C11H14O
1-2,5-dimethylphenyl acetone
CC1=CC(=C(C=C1)C)CC(=O)C
162.232
162.23
≥99.0%
Safety and Handling

Safety and Handling

Recommended Storage : Refrigerator +4°C