missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethylphenylacetone, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC350890050

 View more versions of this product

Catalog No. AC350890050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥99.0%
Liquid
Clear pale yellow
100.0
2, 5-Dimethylphenylacetone
MFCD00210423
Glass bottle
99+%
162.23
5g
99.0
1.5170 to 1.5190 (20°C, 589nm)
C11H14O
53291-89-7
Authentic

Chemical Identifiers

53291-89-7
100.0
C11H14O
MFCD00210423
≥99.0%
99.0
99+%
162.23
2, 5-Dimethylphenylacetone

Safety and Handling

Recommended Storage : Refrigerator +4°C