Please login to your online account to display your discounted pricing

2,5-Dimethylphenylacetone, 99+%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC350890010

 View more versions of this product

Catalog No. AC350890010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 162.23
Quantity 1g
Color Clear pale yellow
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
MDL Number MFCD00210423
CAS 53291-89-7
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 2, 5-Dimethylphenylacetone
Refractive Index 1.5170 to 1.5190 (20°C, 589nm)
Molecular Formula C11H14O
Recommended Storage Refrigerator +4°C

Specifications

Chemical Name or Material 2, 5-Dimethylphenylacetone
CAS 53291-89-7
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C11H14O
Formula Weight 162.23
MDL Number MFCD00210423
Accessories

Refrigerator +4°C