missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethylphenylacetic acid, 98%, ACROS Organics™

Chemical Identifiers

CAS 13612-34-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C10H12O2
Formula Weight 164.2
MDL Number MFCD00014369
Chemical Name or Material 2, 5-Dimethylphenylacetic acid
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC376620050 SDS Fisher Scientific Acros
AC376620050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

13612-34-5
100.0
C10H12O2
MFCD00014369
≥97.5%
97.5
98%
164.2
2, 5-Dimethylphenylacetic acid

Specifications

Crystalline Powder
5g
09, IV, 1850
100.0
97.5
C10H12O2
13612-34-5
2, 5-Dimethylphenylacetic acid
164.2
Light beige
128° to 132°C
MFCD00014369
Glass bottle
Authentic
98%
≥97.5%