Please login to your online account to display your discounted pricing

2,5-Dimethylphenyl isocyanate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 5-Dimethylphenyl isocyanate
Name Note 98%
CAS 40397-98-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9NO
Linear Formula (CH3)C6H3NCO
Formula Weight 147.18
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC366800010

  SDS
thermo fisher scientific
AC366800010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 147.18
Physical Form Liquid
Quantity 1g
Color Clear colorless to slightly yellow
Density 1.043g/mL
CAS Max % 100.0
CAS 40397-98-6
Refractive Index 1.5290 to 1.5310 (20°C, 589nm)
Chemical Name or Material 2, 5-Dimethylphenyl isocyanate
Molecular Formula C9H9NO
Assay Percent Range 98%
CAS Min % 97.5
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Linear Formula (CH3)C6H3NCO
MDL Number MFCD00013854
Recommended Storage Refrigerator +4°C
Name Note 98%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Refrigerator +4°C