missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethylphenoxyacetic acid, 98+%

$65.39 - $211.27

Chemical Identifiers

CAS 7356-41-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00014356
InChI Key RSJMMLSDGNQOEO-UHFFFAOYSA-N
Synonym 2,5-dimethylphenoxyacetic acid, 2,5-dimethylphenoxy acetic acid, 2-2,5-dimethylphenoxy acetic acid, 2,5-xylyloxyacetic acid, 2,5-xylyloxy acetic acid, acetic acid, 2,5-dimethylphenoxy, acetic acid,2-2,5-dimethylphenoxy
PubChem CID 81817
IUPAC Name 2-(2,5-dimethylphenoxy)acetic acid
SMILES CC1=CC(=C(C=C1)C)OCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0117606 View Documents Alfa Aesar™
L0117606
5g Each for $65.39
 
AAL0117614 View Documents Alfa Aesar™
L0117614
25g Each for $211.27
 
Specifications

Chemical Identifiers

7356-41-4
180.203
RSJMMLSDGNQOEO-UHFFFAOYSA-N
81817
CC1=CC(=C(C=C1)C)OCC(=O)O
C10H12O3
MFCD00014356
2,5-dimethylphenoxyacetic acid, 2,5-dimethylphenoxy acetic acid, 2-2,5-dimethylphenoxy acetic acid, 2,5-xylyloxyacetic acid, 2,5-xylyloxy acetic acid, acetic acid, 2,5-dimethylphenoxy, acetic acid,2-2,5-dimethylphenoxy
2-(2,5-dimethylphenoxy)acetic acid

Specifications

114°C to 116°C
7356-41-4
MFCD00014356
2,5-dimethylphenoxyacetic acid, 2,5-dimethylphenoxy acetic acid, 2-2,5-dimethylphenoxy acetic acid, 2,5-xylyloxyacetic acid, 2,5-xylyloxy acetic acid, acetic acid, 2,5-dimethylphenoxy, acetic acid,2-2,5-dimethylphenoxy
CC1=CC(=C(C=C1)C)OCC(=O)O
180.203
180.2
2,5-Dimethylphenoxyacetic acid
5g
C10H12O3
2263621
RSJMMLSDGNQOEO-UHFFFAOYSA-N
2-(2,5-dimethylphenoxy)acetic acid
81817
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning