Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethylphenoxyacetic acid, 98+%


Quantity 25g

Manufacturer: alfa aesar™  L0117614

 View more versions of this product

Catalog No. AAL0117614

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 180.2
Chemical Name or Material 2,5-Dimethylphenoxyacetic acid
Assay ≥98%
CAS 7356-41-4
Molecular Formula C10H12O3
MDL Number MFCD00014356
Beilstein 2263621
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 114° to 116°C
Quantity 25g

Specifications

Chemical Name or Material 2,5-Dimethylphenoxyacetic acid
CAS 7356-41-4
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD00014356
Beilstein 2263621
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.