Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethylphenol, 97%

Chemical Identifiers

Chemical Name or Material 2,5-Dimethylphenol
CAS 95-87-4
Molecular Formula C8H10O
Formula Weight 122.17
MDL Number MFCD00002237
UN Number UN2261
Beilstein 1099260
Merck Index 14,10082
Synonym 2,5-Xylenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA177550B

  SDS
alfa aesar™
A177550B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1775530

  SDS
alfa aesar™
A1775530
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.17
Synonym 2,5-Xylenol
Health Hazard 1 Danger
Merck Index 14,10082
Melting Point 74° to 78°C
MDL Number MFCD00002237
CAS 95-87-4
Quantity 1000g
TSCA TSCA
UN Number UN2261
Flash Point 95°C (203°F)
Molecular Formula C8H10O
Health Hazard 2 GHS H Statement
H310-H314-H318-H302
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Chemical Name or Material 2,5-Dimethylphenol
Beilstein 1099260
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLENOLS, SOLID
Boiling Point 211° to 213°C
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
RTECS Number ZE5775000

Applications
Used to make phenolic resins and (in isomeric mixtures) as an antioxidant for lubricating oils, gasoline, and elastomers. Also used as perfuming agents.

Solubility
Soluble in water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents. Store under dry inert gas.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H310-H314-H318-H302
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.