missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethylphenethylamine, 99%, ACROS Organics™

Chemical Identifiers

CAS 23068-44-2
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD01708273
InChI Key RNEFNTMBBIKOFO-UHFFFAOYSA-N
Synonym 2,5-dimethylphenethylamine
PubChem CID 211415
IUPAC Name 2-(2,5-dimethylphenyl)ethanamine
SMILES CC1=CC(=C(C=C1)C)CCN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC350880010 SDS View Documents Thermo Fisher Scientific
AC350880010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

23068-44-2
149.24
RNEFNTMBBIKOFO-UHFFFAOYSA-N
211415
CC1=CC(=C(C=C1)C)CCN
C10H15N
MFCD01708273
2,5-dimethylphenethylamine
2-(2,5-dimethylphenyl)ethanamine

Specifications

Authentic
1.5300 to 1.532
1g
98.5
98.5% min. (GC)
MFCD01708273
2,5-dimethylphenethylamine
CC1=CC(=C(C=C1)C)CCN
149.24
149.24
99%
Glass bottle
Colorless to Yellow
23068-44-2
100.0
C10H15N
12, IV, 2847
RNEFNTMBBIKOFO-UHFFFAOYSA-N
2-(2,5-dimethylphenyl)ethanamine
211415
Liquid
2, 5-Dimethylphenethylamine
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger