Please login to your online account to display your discounted pricing

2,5-Dimethylphenethylalcohol, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  350870010

 View more versions of this product

Catalog No. AC350870010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 150.22
Quantity 1g
Color Clear colorless
TSCA TSCA
Beilstein 06, IV, 3357
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 2, 5-Dimethylphenethylalcohol
Refractive Index 1.5300 to 1.5320 (20°C, 589nm)
Name Note 99%
Assay Percent Range 99%
Molecular Formula C10 H14 O
Linear Formula (CH3)2C6H3CH2CH2OH
MDL Number MFCD00020610
CAS 6972-51-6
CAS Min % 98.5

Specifications

Chemical Name or Material 2, 5-Dimethylphenethylalcohol
Name Note 99%
CAS 6972-51-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10 H14 O
Linear Formula (CH3)2C6H3CH2CH2OH
Formula Weight 150.22
MDL Number MFCD00020610