Please login to your online account to display your discounted pricing

2,5-Dimethylphenethylalcohol, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 5-Dimethylphenethylalcohol
Name Note 99%
CAS 6972-51-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10 H14 O
Linear Formula (CH3)2C6H3CH2CH2OH
Formula Weight 150.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC350870010

  SDS
fisher scientific acros
350870010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 150.22
Quantity 1g
Color Clear colorless
TSCA TSCA
Beilstein 06, IV, 3357
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 2, 5-Dimethylphenethylalcohol
Refractive Index 1.5300 to 1.5320 (20°C, 589nm)
Name Note 99%
Assay Percent Range 99%
Molecular Formula C10 H14 O
Linear Formula (CH3)2C6H3CH2CH2OH
MDL Number MFCD00020610
CAS 6972-51-6
CAS Min % 98.5