Learn More
2,5-Dimethylhexane, 99%, Thermo Scientific Chemicals
$169.99 - $608.53
Chemical Identifiers
CAS | 592-13-2 |
---|---|
Molecular Formula | C8H18 |
Molecular Weight (g/mol) | 114.23 |
MDL Number | MFCD00008951 |
InChI Key | UWNADWZGEHDQAB-UHFFFAOYSA-N |
Synonym | biisobutyl, hexane, 2,5-dimethyl, diisobutyl, 2,5-dimethyl hexane, hexane,5-dimethyl, acmc-209mbd, 2,5-dimethylhexane, 2,5-dimethylhexane 5g, hexane, 2,5-dimethyl-8ci 9ci |
PubChem CID | 11592 |
IUPAC Name | 2,5-dimethylhexane |
SMILES | CC(C)CCC(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1522106
|
Thermo Scientific Chemicals
A1522106 |
5 g |
Each for $169.99
|
|
|||||
AAA1522114
|
Thermo Scientific Chemicals
A1522114 |
25 g |
Each for $608.53
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
592-13-2 | |
114.23 | |
UWNADWZGEHDQAB-UHFFFAOYSA-N | |
11592 | |
CC(C)CCC(C)C |
C8H18 | |
MFCD00008951 | |
biisobutyl, hexane, 2,5-dimethyl, diisobutyl, 2,5-dimethyl hexane, hexane,5-dimethyl, acmc-209mbd, 2,5-dimethylhexane, 2,5-dimethylhexane 5g, hexane, 2,5-dimethyl-8ci 9ci | |
2,5-dimethylhexane |
Specifications
592-13-2 | |
107°C to 108°C | |
C8H18 | |
MFCD00008951 | |
UN3295 | |
biisobutyl, hexane, 2,5-dimethyl, diisobutyl, 2,5-dimethyl hexane, hexane,5-dimethyl, acmc-209mbd, 2,5-dimethylhexane, 2,5-dimethylhexane 5g, hexane, 2,5-dimethyl-8ci 9ci | |
CC(C)CCC(C)C | |
114.23 | |
114.23 | |
2,5-Dimethylhexane |
0.694 | |
26°C (80°F) | |
1.392 | |
5 g | |
1696877 | |
UWNADWZGEHDQAB-UHFFFAOYSA-N | |
2,5-dimethylhexane | |
11592 | |
99% |
Safety and Handling
GHS H Statement
H304-H226-H315-H336
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Causes skin irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 209-745-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only