Learn More
2,5-Dimethylhexane, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1522114
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,5-Dimethylhexane | |
0.694 | |
26°C (80°F) | |
1.392 | |
25 g | |
1696877 | |
UWNADWZGEHDQAB-UHFFFAOYSA-N | |
2,5-dimethylhexane | |
11592 | |
99% |
592-13-2 | |
107°C to 108°C | |
C8H18 | |
MFCD00008951 | |
UN3295 | |
biisobutyl, hexane, 2,5-dimethyl, diisobutyl, 2,5-dimethyl hexane, hexane,5-dimethyl, acmc-209mbd, 2,5-dimethylhexane, 2,5-dimethylhexane 5g, hexane, 2,5-dimethyl-8ci 9ci | |
CC(C)CCC(C)C | |
114.23 | |
114.23 |
Chemical Identifiers
592-13-2 | |
114.23 | |
UWNADWZGEHDQAB-UHFFFAOYSA-N | |
11592 | |
CC(C)CCC(C)C |
C8H18 | |
MFCD00008951 | |
biisobutyl, hexane, 2,5-dimethyl, diisobutyl, 2,5-dimethyl hexane, hexane,5-dimethyl, acmc-209mbd, 2,5-dimethylhexane, 2,5-dimethylhexane 5g, hexane, 2,5-dimethyl-8ci 9ci | |
2,5-dimethylhexane |
Safety and Handling
GHS H Statement
H304-H226-H315-H336
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Causes skin irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 209-745-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only