Learn More
2,5-Dimethylfuran, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 176030250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2, 5-Dimethylfuran | |
625-86-5 | |
100.0 | |
Colorless to Amber | |
92.0°C to 94.0°C | |
Authentic | |
Glass bottle | |
1.4400 to 1.4420 | |
25 mL | |
0.9 | |
furan, 2,5-dimethyl, 2,5-dimethylfurane, 2,5-dimethyl-furan, 2,5-dimethyl furane, unii-dr5hl9oj7y, ccris 3158, 2,5-dimethyl furan, dr5hl9oj7y, 2.5-dimethyl furan, acmc-1avpg | |
GSNUFIFRDBKVIE-UHFFFAOYSA-N | |
2,5-dimethylfuran | |
12266 | |
99% |
99% | |
98.5 | |
-62.0°C | |
0.9000g/mL | |
−1°C | |
98.5% min. (GC) | |
C6H8O | |
MFCD00003250 | |
17,41 | |
15,3268 | |
Solubility in water: insoluble. Other solubilities: soluble in oil,organic solvents | |
CC1=CC=C(O1)C | |
96.13 | |
96.13 | |
Liquid |
Chemical Identifiers
625-86-5 | |
96.13 | |
GSNUFIFRDBKVIE-UHFFFAOYSA-N | |
12266 | |
CC1=CC=C(O1)C |
C6H8O | |
MFCD00003250 | |
furan, 2,5-dimethyl, 2,5-dimethylfurane, 2,5-dimethyl-furan, 2,5-dimethyl furane, unii-dr5hl9oj7y, ccris 3158, 2,5-dimethyl furan, dr5hl9oj7y, 2.5-dimethyl furan, acmc-1avpg | |
2,5-dimethylfuran |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Highly flammable liquid and vapour.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Danger
EINECSNumber : 210-914-3
RTECSNumber : LU0875000
TSCA : TSCA
Recommended Storage : May darken on storage
RUO – Research Use Only