missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethylcinnamic acid, 98+%

$110.57 - $382.45

Chemical Identifiers

CAS 95883-10-6
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00014377
InChI Key FAIBVLSPNAHVSQ-UHFFFAOYSA-N
Synonym 3-2,5-dimethylphenyl prop-2-enoic acid, 2,5-dimethylcinnamic acid, 2-propenoic acid, 3-2,5-dimethylphenyl
PubChem CID 53401582
IUPAC Name 3-(2,5-dimethylphenyl)prop-2-enoic acid
SMILES CC1=CC(=C(C=C1)C)C=CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0179904 View Documents Alfa Aesar™
L0179904
2g Each for $110.57
 
AAL0179909 View Documents Alfa Aesar™
L0179909
10g Each for $382.45
 
Description

Description

Applications
2,5-Dimethylcinnamic acid, is used for organic synthesize intermediate

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

95883-10-6
176.215
FAIBVLSPNAHVSQ-UHFFFAOYSA-N
53401582
CC1=CC(=C(C=C1)C)C=CC(=O)O
C11H12O2
MFCD00014377
3-2,5-dimethylphenyl prop-2-enoic acid, 2,5-dimethylcinnamic acid, 2-propenoic acid, 3-2,5-dimethylphenyl
3-(2,5-dimethylphenyl)prop-2-enoic acid

Specifications

129°C to 131°C
95883-10-6
MFCD00014377
3-2,5-dimethylphenyl prop-2-enoic acid, 2,5-dimethylcinnamic acid, 2-propenoic acid, 3-2,5-dimethylphenyl
CC1=CC(=C(C=C1)C)C=CC(=O)O
176.215
176.22
2,5-Dimethylcinnamic acid
2g
C11H12O2
3244516
FAIBVLSPNAHVSQ-UHFFFAOYSA-N
3-(2,5-dimethylphenyl)prop-2-enoic acid
53401582
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning