Please login to your online account to display your discounted pricing

2,5-Dimethylbenzylamine, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC350860010

 View more versions of this product

Catalog No. AC350860010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 135.21
Quantity 1g
Color Clear colorless
CAS Max % 100.0
Chemical Name or Material 2, 5-Dimethylbenzylamine
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 98%
Molecular Formula C9H13N
MDL Number MFCD00039780
CAS 93-48-1
CAS Min % 98.5
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.5350 to 1.5380 (20°C, 589nm)

Specifications

Chemical Name or Material 2, 5-Dimethylbenzylamine
Name Note 99%
CAS 93-48-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H13N
Formula Weight 135.21
MDL Number MFCD00039780
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents