missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethylbenzyl Chloride 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 155290250

 View more versions of this product

Catalog No. AC155290250


 

Specifications Description & Specifications

Specifications

2, 5-Dimethylbenzyl chloride
Authentic
25g
824-45-3
100.0
1.5364 to 1.5384 (20°C, 589nm)
≥97.5%
C9 H11 Cl
154.64
05, I, 195
1.0400g/mL
Clear colorless
98%
97.5
Glass bottle
Liquid
98%
(CH3)2C6H3CH2Cl
MFCD00000902
1-Chloromethyl-2, 5-dimethylbenzene

Chemical Identifiers

824-45-3
100.0
C9 H11 Cl
154.64
1-Chloromethyl-2, 5-dimethylbenzene
≥97.5%
97.5
98%
(CH3)2C6H3CH2Cl
MFCD00000902
2, 5-Dimethylbenzyl chloride, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 212-529-6

TSCA : TSCA