Please login to your online account to display your discounted pricing

2,5-Dimethylbenzyl Chloride 98%, ACROS Organics™

$119.50 - $119.50

Chemical Identifiers

Chemical Name or Material 2, 5-Dimethylbenzyl chloride
Name Note 98%
CAS 824-45-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9 H11 Cl
Linear Formula (CH3)2C6H3CH2Cl
Formula Weight 154.64
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC155290250

  SDS
thermo fisher scientific
155290250
Glass bottle 25g Each for $119.50
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 154.64
Physical Form Liquid
Quantity 25g
Color Clear colorless
Beilstein 05, I, 195
TSCA TSCA
EINECS Number 212-529-6
Density 1.0400g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Refractive Index 1.5364 to 1.5384 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
CAS Min % 97.5
Synonym 1-Chloromethyl-2, 5-dimethylbenzene
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 2, 5-Dimethylbenzyl chloride
Name Note 98%
Molecular Formula C9 H11 Cl
MDL Number MFCD00000902
CAS 824-45-3
Assay Percent Range 98%
Linear Formula (CH3)2C6H3CH2Cl