Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethylbenzoxazole, 99%

$46.80 - $110.82

Chemical Identifiers

Chemical Name or Material 2,5-Dimethylbenzoxazole
CAS 5676-58-4
Molecular Formula C9H9NO
Formula Weight 147.18
MDL Number MFCD00005773
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2178003

  SDS
alfa aesar™
B2178003
1g Each for $46.80

AAB2178006

  SDS
alfa aesar™
B2178006
5g Each for $110.82
Description & Specifications

Specifications

Formula Weight 147.18
Chemical Name or Material 2,5-Dimethylbenzoxazole
TSCA TSCA
Assay 99%
Molecular Formula C9H9NO
MDL Number MFCD00005773
CAS 5676-58-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 218° to 219°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.