Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethylbenzothiazole, 99%

$50.18 - $167.78

Chemical Identifiers

Chemical Name or Material 2,5-Dimethylbenzothiazole
CAS 95-26-1
Molecular Formula C9H9NS
Formula Weight 163.24
MDL Number MFCD00005796
Beilstein 116455
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1483006

 
alfa aesar™
A1483006
5g Each for $50.18

AAA1483014

 
alfa aesar™
A1483014
25g Each for $167.78
Description & Specifications

Specifications

Formula Weight 163.24
Chemical Name or Material 2,5-Dimethylbenzothiazole
TSCA TSCA
Assay 99%
Molecular Formula C9H9NS
Melting Point 37° to 42°C
MDL Number MFCD00005796
CAS 95-26-1
Beilstein 116455
Quantity 5g
Flash Point >110°C (230°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DL3675000

Applications
2,5-Dimethylbenzothiazole is used as intermediates for medicines and materials, chemical reagents and fine chemicals.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.