Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethylbenzoic acid, 96%

$73.43 - $240.12

Chemical Identifiers

Chemical Name or Material 2,5-Dimethylbenzoic acid
CAS 610-72-0
Molecular Formula C9H10O2
Formula Weight 150.18
MDL Number MFCD00002482
Beilstein 971588
Synonym p-Xylylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1535209

 
alfa aesar™
A1535209
10g Each for $73.43

AAA1535218

 
alfa aesar™
A1535218
50g Each for $240.12
Description & Specifications

Specifications

Formula Weight 150.18
Chemical Name or Material 2,5-Dimethylbenzoic acid
TSCA TSCA
Assay 96%
Molecular Formula C9H10O2
Melting Point 132° to 135°C
Boiling Point 268°C (subl.)
MDL Number MFCD00002482
CAS 610-72-0
Beilstein 971588
Quantity 10g
Odor Odorless
Synonym p-Xylylic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
2,5-Dimethylbenzoic acid acts as a metabolite of pseudocumene. 2,5-Dimethylbenzoic acid was used in the synthesis of new derivatives of [24]paracyclophane. It was used as starting reagent in the synthesis of methyl 2,5-dimethylbenzoate.

Solubility
Solubility - Very faint turbidity with Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.