missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethylbenzenesulfonyl chloride, 98%

Manufacturer:  Alfa Aesar™ 4482606

 View more versions of this product

Catalog No. AA4482606



Specifications

Specifications

2,5-Dimethylbenzenesulfonyl chloride
120°C to 125°C
20°C
19040-62-1
MFCD00024875
p-xylene-2-sulfonyl chloride, benzenesulfonyl chloride, 2,5-dimethyl, 2,5-dimethylbenzene-1-sulfonyl chloride, 2,5-dimethylphenylsulfonyl chloride, 2,5-dimethyl-benzenesulfonyl chloride, benzenesulfonylchloride, 2,5-dimethyl, 2,5-dimethylphenyl chlorosulfone, acmc-209etq, xylenesulfonylchloride, ksc178m0f
CC1=CC(=C(C=C1)C)S(=O)(=O)Cl
204.668
204.68
98%
Moisture Sensitive
Yellow
5g
C8H9ClO2S
UN3265
FZVZUIBYLZZOEW-UHFFFAOYSA-N
2,5-dimethylbenzenesulfonyl chloride
87910
Liquid/Solid

Chemical Identifiers

19040-62-1
204.668
FZVZUIBYLZZOEW-UHFFFAOYSA-N
87910
CC1=CC(=C(C=C1)C)S(=O)(=O)Cl
C8H9ClO2S
MFCD00024875
p-xylene-2-sulfonyl chloride, benzenesulfonyl chloride, 2,5-dimethyl, 2,5-dimethylbenzene-1-sulfonyl chloride, 2,5-dimethylphenylsulfonyl chloride, 2,5-dimethyl-benzenesulfonyl chloride, benzenesulfonylchloride, 2,5-dimethyl, 2,5-dimethylphenyl chlorosulfone, acmc-209etq, xylenesulfonylchloride, ksc178m0f
2,5-dimethylbenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : DB8927600

TSCA : TSCA

SDS Documents