Please login to your online account to display your discounted pricing

2,5-Dimethylbenzenesulfonyl chloride, 97%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  MO07001EA

 View more versions of this product

Catalog No. MO07001EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 204.68
Quantity 10g
Assay 97%
CAS Min % 97.0
Linear Formula (CH3)2C6H3SO2Cl
Chemical Name or Material 2, 5-Dimethylbenzenesulfonyl chloride
Assay Percent Range 97%
Molecular Formula C8H9ClO2S
MDL Number MFCD00024875
CAS 19040-62-1
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 5-Dimethylbenzenesulfonyl chloride
CAS 19040-62-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H9ClO2S
Linear Formula (CH3)2C6H3SO2Cl
Formula Weight 204.68
MDL Number MFCD00024875
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents