Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethylanisole, 98+%

$71.72 - $182.32

Chemical Identifiers

Chemical Name or Material 2,5-Dimethylanisole
CAS 1706-11-2
Molecular Formula C9H12O
Formula Weight 136.19
MDL Number MFCD00008378
Beilstein 774604
Synonym 2-Methoxy-p-xylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2045614

  SDS
alfa aesar™
B2045614
25g Each for $71.72

AAB2045622

  SDS
alfa aesar™
B2045622
100g Each for $182.32
Description & Specifications

Specifications

Formula Weight 136.19
Quantity 25g
Flash Point 66°C (150°F)
Chemical Name or Material 2,5-Dimethylanisole
Beilstein 774604
Synonym 2-Methoxy-p-xylene
Health Hazard 1 Warning
MDL Number MFCD00008378
Assay ≥98%
Molecular Formula C9H12O
Boiling Point 189° to 190°C
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
CAS 1706-11-2
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.