Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethylaniline, 98+%

Chemical Identifiers

Chemical Name or Material 2,5-Dimethylaniline
CAS 95-78-3
Molecular Formula C8H11N
Formula Weight 121.18
MDL Number MFCD00007743
UN Number UN1711
Beilstein 2205178
Merck Index 14,10084
Synonym 2,5-Xylidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1268236

  SDS
alfa aesar™
A1268236
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 121.18
Chemical Name or Material 2,5-Dimethylaniline
TSCA TSCA
Assay ≥98%
Molecular Formula C8H11N
MDL Number MFCD00007743
CAS 95-78-3
UN Number UN1711
Beilstein 2205178
Merck Index 14,10084
Synonym 2,5-Xylidine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280h-P273-P309-P310
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLIDINES, LIQUID
RTECS Number ZE9100000
Flash Point 93°C (199°F)
Odor Characteristic
Boiling Point 218°C
Melting Point 15°C
Quantity 500g

Applications
2,5-Dimethylaniline is widely used as raw materials to produce imaging chemicals like organic dyes and pigments. It is also used in the production of antioxidants, pharmaceutical, agricultural, rubber chemicals and other target organic molecules. 2,5-Dimethylaniline was also used in the synthesis of nanocomposite of multi-walled carbon nanotubes embedded in poly(2,5-dimethylaniline). 2,5-Dimethylaniline was used to study the effect of metabolites formed from 2,5-xylidine by fungi on laccase activity. The Fourier transform infrared (FTIR) and Raman spectra of 2,5-dimethylaniline was studied5. It induced transcription of lccIV.

Solubility
Soluble in water <0.1 g/100 mL at 18°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280h-P273-P309-P310
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.