Please login to your online account to display your discounted pricing

2,5-Dimethyl-p-phenylenediamine, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC377050010

 View more versions of this product

Catalog No. AC377050010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 136.2
Color Grayish-brown
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Linear Formula (CH3)2C6H2(NH2)2
Chemical Name or Material 2, 5-Dimethyl-p-phenylenediamine
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C8H12N2
MDL Number MFCD00052675
CAS 6393-01-7
Synonym 2, 5-Diamino-p-xylene
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2, 5-Dimethyl-p-phenylenediamine
CAS 6393-01-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H12N2
Linear Formula (CH3)2C6H2(NH2)2
Formula Weight 136.2
MDL Number MFCD00052675
Synonym 2, 5-Diamino-p-xylene
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents