missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethyl-p-phenylenediamine, 97%, ACROS Organics™

Chemical Identifiers

CAS 6393-01-7
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H12N2
Linear Formula (CH3)2C6H2(NH2)2
Formula Weight 136.2
MDL Number MFCD00052675
Synonym 2, 5-Diamino-p-xylene
Chemical Name or Material 2, 5-Dimethyl-p-phenylenediamine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC377050010 SDS Thermo Fisher Scientific
AC377050010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6393-01-7
100.0
C8H12N2
136.2
2, 5-Diamino-p-xylene
≥96.0%
96.0
97%
(CH3)2C6H2(NH2)2
MFCD00052675
2, 5-Dimethyl-p-phenylenediamine

Specifications

136.2
Crystalline Powder
96.0
(CH3)2C6H2(NH2)2
97%
6393-01-7
2, 5-Diamino-p-xylene
2, 5-Dimethyl-p-phenylenediamine
Grayish-brown
1g
100.0
Authentic
MFCD00052675
Glass bottle
C8H12N2
≥96.0%

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning