missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,5-Dimethyl-p-benzoquinone, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC408230250
Specifications
2, 5-Dimethyl-p-benzoquinone | |
Glass bottle | |
123°C to 126°C | |
137-18-8 | |
100.0 | |
C8H8O2 | |
2,5-cyclohexadiene-1,4-dione, 2,5-dimethyl, 2,5-dimethyl-1,4-benzoquinone, 2,5-dimethyl-p-benzoquinone, 2,5-dimethylquinone, 2,5-xyloquinone, 3,6-dimethyl-p-benzoquinone, floron, floron czech, p-xyloquinone, phlorone | |
CC1=CC(=O)C(=CC1=O)C | |
136.15 | |
136.15 | |
99% |
Authentic | |
Brown to Yellow | |
25g | |
98.5 | |
98.5% min. (GC) | |
MFCD00041737 | |
MYKLQMNSFPAPLZ-UHFFFAOYSA-N | |
2,5-dimethylcyclohexa-2,5-diene-1,4-dione | |
8718 | |
Crystalline Powder |
Chemical Identifiers
137-18-8 | |
136.15 | |
MYKLQMNSFPAPLZ-UHFFFAOYSA-N | |
8718 | |
CC1=CC(=O)C(=CC1=O)C |
C8H8O2 | |
MFCD00041737 | |
2,5-cyclohexadiene-1,4-dione, 2,5-dimethyl, 2,5-dimethyl-1,4-benzoquinone, 2,5-dimethyl-p-benzoquinone, 2,5-dimethylquinone, 2,5-xyloquinone, 3,6-dimethyl-p-benzoquinone, floron, floron czech, p-xyloquinone, phlorone | |
2,5-dimethylcyclohexa-2,5-diene-1,4-dione |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep a
GHS Signal Word: Danger
EINECSNumber : 205-283-6