Learn More
2,5-Dimethyl-p-benzoquinone, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 408230050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 5-Dimethyl-p-benzoquinone | |
98.5 | |
Authentic | |
Glass bottle | |
MFCD00041737 | |
2,5-dimethyl-1,4-benzoquinone, phlorone, p-xyloquinone, 2,5-dimethyl-p-benzoquinone, 2,5-xyloquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dimethyl, 2,5-dimethylquinone, floron, 3,6-dimethyl-p-benzoquinone, floron czech | |
CC1=CC(=O)C(=CC1=O)C | |
136.15 | |
136.15 |
137-18-8 | |
100.0 | |
98.5% min. (GC) | |
C8H8O2 | |
5g | |
MYKLQMNSFPAPLZ-UHFFFAOYSA-N | |
2,5-dimethylcyclohexa-2,5-diene-1,4-dione | |
8718 | |
99% |
Chemical Identifiers
137-18-8 | |
136.15 | |
MYKLQMNSFPAPLZ-UHFFFAOYSA-N | |
8718 | |
CC1=CC(=O)C(=CC1=O)C |
C8H8O2 | |
MFCD00041737 | |
2,5-dimethyl-1,4-benzoquinone, phlorone, p-xyloquinone, 2,5-dimethyl-p-benzoquinone, 2,5-xyloquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dimethyl, 2,5-dimethylquinone, floron, 3,6-dimethyl-p-benzoquinone, floron czech | |
2,5-dimethylcyclohexa-2,5-diene-1,4-dione |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep a
GHS Signal Word: Danger
EINECSNumber : 205-283-6
RUO â Research Use Only