Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dimethyl-4-(4-morpholinylmethyl)phenol hydrochloride hydrate, 99%

$22.91 - $74.58

Chemical Identifiers

Chemical Name or Material 2,5-Dimethyl-4-(4-morpholinylmethyl)phenol hydrochloride hydrate
CAS 33625-43-3
Molecular Formula C13H19NO2·HCl·xH2O
Formula Weight 257.76 (Anhydrous)
MDL Number MFCD00150097
Beilstein 4024300
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0402414

  SDS
alfa aesar™
L0402414
25g Each for $22.91

AAL0402422

  SDS
alfa aesar™
L0402422
100g Each for $74.58
Description & Specifications

Specifications

Formula Weight 257.76 (Anhydrous)
CAS 33625-43-3
MDL Number MFCD00150097
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2,5-Dimethyl-4-(4-morpholinylmethyl)phenol hydrochloride hydrate
Assay 99%
Molecular Formula C13H19NO2·HCl·xH2O
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
Health Hazard 1 Warning
Melting Point 181° to 185°C
Quantity 25g
Beilstein 4024300
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.