missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethyl-3-hexyne-2,5-diol, (+/-) + meso, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2088936

 View more versions of this product

Catalog No. AAB2088936



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,5-Dimethyl-3-hexyne-2,5-diol
142-30-3
0.949
102°C (215°F)
MFCD00004468
969835
IHJUECRFYCQBMW-UHFFFAOYSA-N
2,5-dimethylhex-3-yne-2,5-diol
8883
98%
(+/-) + meso
90°C to 94°C
205°C
C8H14O2
500 g
2,5-dimethyl-3-hexyne-2,5-diol, dimethylhexynediol, kemitracin-50, tetramethylbutynediol, acetylenepinacol, 3-hexyne-2,5-diol, 2,5-dimethyl, olfine y, tetramethyl-2-butynediol, unii-22rr53u71w, 2,5-dimethylhexyne-2,5-diol
CC(C)(C#CC(C)(C)O)O
142.198
142.2

Chemical Identifiers

142-30-3
142.198
IHJUECRFYCQBMW-UHFFFAOYSA-N
8883
CC(C)(C#CC(C)(C)O)O
C8H14O2
MFCD00004468
2,5-dimethyl-3-hexyne-2,5-diol, dimethylhexynediol, kemitracin-50, tetramethylbutynediol, acetylenepinacol, 3-hexyne-2,5-diol, 2,5-dimethyl, olfine y, tetramethyl-2-butynediol, unii-22rr53u71w, 2,5-dimethylhexyne-2,5-diol
2,5-dimethylhex-3-yne-2,5-diol
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 205-533-4

RTECSNumber : MR0176000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only